Would you like to log in again?
VWR® C-Leg Lab Benches, Frame + Top + Uprights + Shelves, Motor, Snow White
Supplier: Avantor
VWR®
14-C103SW429 14-C103SW430 14-C103SW475 14-C100SW466 14-C103SW423 14-C100SW465 14-C103SW460 14-C103SW420 14-C103SW466 14-C103SW436 14-C103SW474 14-C103SW471 14-C100SW473 14-C100SW472 14-C103SW470 14-C100SW461 14-C100SW457 14-C103SW421 14-C100SW470 14-C103SW426 14-C100SW469 14-C103SW467 14-C100SW474 14-C100SW463 14-C103SW427 14-C103SW468 14-C103SW462 14-C103SW432 14-C103SW464 14-C103SW439 14-C100SW458 14-C100SW459 14-C103SW428 14-C103SW457 14-C100SW467 14-C100SW464 14-C100SW462 14-C100SW475 14-C103SW472 14-C103SW422 14-C100SW460 14-C103SW463 14-C103SW473 14-C103SW435 14-C103SW458 14-C103SW433 14-C100SW471 14-C103SW431 14-C100SW456 14-C103SW465 14-C103SW434 14-C103SW461 14-C103SW437 14-C103SW459 14-C103SW456 14-C103SW424 14-C103SW425 14-C100SW468 14-C103SW469 14-C103SW438
76551-074EA 2542.37 USD InStock
76551-074 76551-072 76551-194 76550-904 76551-036 76550-996 76550-710 76550-832 76550-316 76550-834 76550-758 76550-714 76550-470 76550-396 76551-166 76551-122 76551-002 76551-120 76551-042 76550-914 76551-008 76551-004 76550-884 76550-400 76550-642 76550-604 76550-924 76550-684 76550-366 76551-178 76551-172 76550-808 76550-698 76550-654 76550-776 76550-898 76550-932 76550-570 76550-890 76550-572 76550-772 76551-186 76551-140 76550-380 76551-108 76551-102 76550-620 76550-864 76550-502 76550-546 76550-900 76550-306 76550-946 76550-748 76551-060 76550-584 76550-464 76550-662 76550-784 76550-344
VWR® C-Leg Lab Benches, Frame + Top + Uprights + Shelves, Motor, Snow White
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel. There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality.
  • Snow white frame
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Offering a wide range of accessories to meet any need
  • Various concept workbench sizes and combinations
  • Height adjustment available either electrically (electric motor height adjustment) or with a hand crank or an allen key
  • A wide range of accessories and add-ons, making concept workbenches easy to customize
  • Impressive stability with its steel frame and maximum load capacity of 1100 lbs.
  • Cleanroom suitability ISO class 7
The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Shelves depth 11.81".
Accessories information: Upright accessories can be added in any position along the upright in 1" increments. Some accessories are bench style or size dependent.
Ordering information: Manual, hand crank, and motorized frame styles are available, as well as several configurations, including frame only, frame + top, frame + top + uprights, frame + top + uprights + shelves, and frames with notches for uprights (all sold separately). Contact your Avantor representative for more information. Notched work surfaces add 2.625" to depth of table frame. Work surfaces for single bay uprights include 2 notches (left and right); work surfaces for double bay uprights include 3 notches (left, center, right). Tables ship LTL, flat-packed. Installation not included.
Packaging: Benches are shipped knocked down, on non-stackable pallets. Assembly instructions and hardware included.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty