Would you like to log in again?
Ohmeda Panda 3300 Infant Warmer
Supplier: GUMBO MEDICAL
OP3300
470346-576EA 3272.69 USD InStock
470346-576
Ohmeda Panda 3300 Infant Warmer
Incubators Infant Incubators
Warming for labor and delivery.
  • Refurbished
  • 1 year parts warranty
  • For educational and training use only
The Ohmeda Medical Panda™ Model 3300 Baby Warmer System provides a controlled source of radiant heat for infant and pediatric patients. The control system uses a microprocessor and provides both manual and servo modes of operation. The patient temperature, control temperature, and elapsed time displays are digital for ease of viewing. The control panel includes a one-hour elapsed timer with time displayed in minutes and seconds. The timer features optional audible tones for use during Apgar scoring. A complete audio and visual alarm system is included on the control panel. The model 3300 has integral bed for infants and is intended for procedures and long-term care in the Nursery and Labor and Delivery.
Delivery information: This item has to ship by freight, Additional shipping charges may apply.
Caution: This product is refurbished and is intended for educational and training use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty