Would you like to log in again?
Klerwipe™ non-woven dry wipes
Supplier: Ecolab
Klerwipe™
53000344 53000346
76554-280CS 1050.65 USD InStock
76554-280 76554-278
Klerwipe™ non-woven dry wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Klerwipe™ Non-Woven Dry Wipes are low particulate wipes for cleanrooms.
  • Autoclavable
  • High absorbency and durable
  • Chemically compatible with disinfectants and cleaning solutions
  • Nonwoven dry wipers are more absorbent than knitted wipes
  • Binder free means no residue are left on critical surfaces
  • An excellent value combined with high performance cleaning characteristics
Klerwipe™ Non-Woven Dry Wipes are general purpose cleanroom wipes. Made from a binder free 55% cellulose/ 45% polyester hydroentangled nonwoven blend.
Recommended for use in ISO Class 5 and above environments.
Packaging: Double bagged.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty