Would you like to log in again?
Klerwipe™ knitted dry wipes
Supplier: Ecolab
Klerwipe™
53000349 53000352
76554-282CS 884.76 USD InStock
76554-282 76554-284
Klerwipe™ knitted dry wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Klerwipe™ knitted dry wipes are low particulate wipes for cleanrooms.
  • Soft hand and edge for sensitive surfaces, abrasion resistant
  • Low particle and fiber generation, high level of cleanliness
  • Chemically compatible with disinfectants and cleaning solutions
  • Sealed edge, cleanroom laundered, knitted wipes offer higher purity and durability compared to nonwoven wipes
Klerwipe™ Knitted Dry Wipes are general purpose cleanroom wipe. Ultrasonically sealed edge wipe is 100% knitted polyester and laundered and packaged in an ISO Class 4 cleanroom.
Recommended for use in ISO Class 3 and above environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty