Would you like to log in again?
Skolnik UN Certified Steel Open Head Drums with Lever lock Ring Closure, Qorpak®
Supplier: Qorpak
1721- 1653- 1521-
76579-190EA 234.66 USD InStock
76579-190 76579-180 76579-182
Skolnik UN Certified Steel Open Head Drums with Lever lock Ring Closure, Qorpak®
Barrels, Kegs, Drums
Qorpak® Skolnik UN certified steel drums are thicker, heavier, and stronger than industry standards require, offer extreme durability, and comply with domestic and international regulations.
  • Meets the rigorous UN packaging requirements of 49 CFR
  • Tested per the DOT and UN packaging test requirements
  • Heavier and stronger than industry standards
  • Better endures nail and forklift punctures than thinner drums
Do not risk place your volatile, expensive, irreplaceable, hazardous, or precious contents in any other vessel. Protect your products, reputation, and the environment with a superior quality drum.
Certifications: UN certified for solids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty