Would you like to log in again?
Pluronic® F-127 Cell culture tested
Supplier: AAT Bioquest
Pluronic®
20050
76482-986EA 93.54 USD InStock
76482-986
Pluronic® F-127 Cell culture tested
Cleaning Agents Detergents and Surface Cleansers
Pluronic® F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility.
  • Non-toxic to cells
It is soluble at low temperature, but gels at room temperature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty