Would you like to log in again?
Alcohol Prep Pads, DUKAL™ Corporation
Supplier: Dukal
853-400M
76419-816CS 88.39 USD InStock
76419-816
Alcohol Prep Pads, DUKAL™ Corporation
Wipes Disposable Wipes and Towels Disinfectant Wipes
These sterile pads are saturated with 70% Isopropyl Alcohol to cleanse minor cuts and abrasions.
  • Individually sealed to ensure that they remain fully saturated
  • 2-ply, non-woven pads
  • Without natural rubber latex
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty