Would you like to log in again?
SatPax® MicroSeal®-VP Pre-wetted Sealed Edge, 100% Polyester Knit, 60% Saturation, Berkshire
Supplier: Berkshire
SatPax®
SPXVPR00112
76552-176CS 321.82 USD InStock
76552-176
SatPax® MicroSeal®-VP Pre-wetted Sealed Edge, 100% Polyester Knit, 60% Saturation, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SatPax® MicroSeal®-VP is an ultrasonically sealed edge cleanroom laundered wiper recommended for ISO Class 3 and above environments composed of 100% continuous filament polyester knit fabric.
  • Pre-wetted with 70% IPA, 30% DI water, 60% saturation
  • 100% continuous filament polyester knit
  • Ultrasonically sealed edge for reduced fiber contamination
  • Laundered and packaged in Berkshire’s ISO Class 4 cleanroom with lot number on each pack
  • Lightweight, high absorbency material design reduces actual cost in use compared to heavier weight products
  • Critically low particles, fibers, ions and extractables
SatPax® MicroSeal®-VP is an ultrasonically sealed edge cleanroom laundered wiper recommended for ISO Class 3 and above environments composed of 100% continuous filament polyester knit fabric. The wiper is pre-wetted with a consistent concentration of IPA and DI water and a 100% IPA option. This combination of properties provides SatPax® MicroSeal ®-VP with the highest cleanliness, abrasion resistance, and chemical compatibility required for cleaning critical environments. The Pre-wetted format offers a cost-effective and easy-to-use solution versus traditional bulk handling of solvents and maintenance of squirt bottles.
Certifications: ISO Class 3 – 5, Cleanroom Class 1 – 100. EU Grade B/C/D.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty