Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Europium(III) oxide 99.99%

Supplier: APOLLO SCIENTIFIC
Warning

IN1789-5G IN1789-25G
76563-178EA 189 USD InStock
76563-178 76560-770
Europium(III) oxide 99.99%
Europium (III) oxide


Formula: Eu₂O₃
MW: 351.92 g/mol
Boiling Pt: 4118 °C
Melting Pt: 2291 °C
Density: 7.42 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010997
CAS Number: 1308-96-9
EINECS: 215-165-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty