Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiophosphate hydrate

Supplier: APOLLO SCIENTIFIC
Warning

IN3322-1G IN3322-5G IN3322-25G
76562-814EA 221 USD InStock
76562-814 76560-146 76560-774
Sodium thiophosphate hydrate
Sodium thiophosphate hydrate


Formula: H₂Na₃O₄PS
Melting Pt: 56…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041513
CAS Number: 10489-48-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty