Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium(II) acetate 99.99+%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Palladium diacetate

IN3837-5G IN3837-1G
76560-702EA 530 USD InStock
76560-702 76561-114
Palladium(II) acetate 99.99+%
Palladium (II) acetate


Formula: C₄H₆O₄Pd
MW: 224.51 g/mol
Melting Pt: 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012453
CAS Number: 3375-31-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty