Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tungstate dihydrate 98-101% ACS

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Sodium tungsten oxide dihydrateTungstic acid sodium salt dihydrateSodium wolframate dihydrate

IN3275-100G IN3275-1KG
76561-868EA 63 USD InStock
76561-868 76559-594
Sodium tungstate dihydrate 98-101% ACS
Sodium wolframate dihydrate


Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Melting Pt: 698 °C
Density: 3.23-3.25 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2
EINECS: 236-743-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty