Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diyttrium trioxide 99,9%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Yttrium (III) oxideYttrium oxide

IN3862-25G
76561-066EA 14.35 USD InStock
76561-066
Diyttrium trioxide 99,9%
Diyttrium trioxide


Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300 °C
Melting Pt: 2410 °C
Density: 4.84 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty