Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-chlorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Bromochlorobenzene

OR0017-25G OR0017-250G OR0017-100G
76559-774EA 70 USD InStock
76559-774 76562-526 76559-596
1-Bromo-2-chlorobenzene 99%
1-Bromo-2-chlorobenzene


Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 203…205 °C
Melting Pt: –13 °C
Density: 1.649 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000532
CAS Number: 694-80-4
EINECS: 211-775-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty