Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Acetyl-2-naphthol 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-(2-Hydroxy-1-naphthyl)ethan-1-one2'-Hydroxy-1'-acetonaphthone1-Acetyl-2-hydroxynaphthalene1-(2-Hydroxynaphthalen-1-yl)ethanone2-Hydroxy-1-acetonaphthone

OR015967-100G OR015967-25G
76560-814EA 52 USD InStock
76560-814 76560-546
1-Acetyl-2-naphthol 97%
1-Acetyl-2-naphthol


Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 130…132 °C/2mmHg
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00012131
CAS Number: 574-19-6
EINECS: 209-369-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty