Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitroindole 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-Nitro-1H-indole

OR0385-500G OR0385-25G OR0385-100G
76560-804EA 82 USD InStock
76560-804 76560-124 76559-902
5-Nitroindole 98%
5-Nitroindole


Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 139…143 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005673
CAS Number: 6146-52-7
EINECS: 228-153-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty