Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxy-4-nitrobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR0535-25G OR0535-100G OR0535-500G
76561-528EA 13.33 USD InStock
76561-528 76560-976 76560-252
3-Hydroxy-4-nitrobenzoic acid
3-Hydroxy-4-nitrobenzoic acid


Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 229…236 °C
Storage Temperature: Ambient
MDL Number: MFCD00007110
CAS Number: 619-14-7
EINECS: 210-580-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty