Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodo-2-nitrotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0542-25G OR0542-5G OR0542-100G
76562-626EA 16 USD InStock
76562-626 76561-098 76561-580
4-Iodo-2-nitrotoluene 98%
4-Iodo-2-nitrotoluene


Formula: C₇H₆INO₂
MW: 263.03 g/mol
Melting Pt: 66…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00051090
CAS Number: 41252-97-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty