Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-α-Phenylalanine 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Phenyl-D(+)-alanine3-Phenyl-D-alanineH-D-Phe-OHD-Phenylalanine

OR0702-5G
76561-244EA 51 USD InStock
76561-244
D-α-Phenylalanine 97%
D-α-Phenylalanine


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 273…276 °C
Storage Temperature: Ambient
MDL Number: MFCD00004270
CAS Number: 673-06-3
EINECS: 211-603-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty