Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(2-chloro-4-nitrophenyl)piperazine 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-(2-Chloro-4-nitrophenyl)piperazine

OR0871-5G OR0871-25G OR0871-1G
76561-934EA 134 USD InStock
76561-934 76560-232 76561-948
1-(2-chloro-4-nitrophenyl)piperazine 95%
1-(2-chloro-4-nitrophenyl)piperazine


Formula: C₁₀H₁₂ClN₃O₂
MW: 241.675 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00640776
CAS Number: 114878-60-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty