Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-bromo-3-methylbenzoate

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Methyl 4-bromo-3-methylbenzoate

OR0924-1G OR0924-25G OR0924-5G
76561-758EA 16 USD InStock
76561-758 76561-834 76559-840
Methyl-4-bromo-3-methylbenzoate
Methyl 4-bromo-3-methylbenzoate


Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Melting Pt: 38…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00673014
CAS Number: 148547-19-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty