Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylammonium iodide

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: TBAITetra-n-butylammonium iodide

OR10056-100G OR10056-25G OR10056-5G
76560-280EA 16 USD InStock
76560-280 76561-972 76562-222
Tetrabutylammonium iodide
Tetrabutylammonium iodide


Formula: C₁₆H₃₆IN
MW: 369.37 g/mol
Melting Pt: 142…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00011636
CAS Number: 311-28-4
EINECS: 206-220-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty