Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(3-Bromopropyl)benzene 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Bromo-3-phenylpropane

OR10084-1G OR10084-5G OR10084-25G
76560-378EA 13.33 USD InStock
76560-378 76559-490 76560-330
(3-Bromopropyl)benzene 95%
(3-Bromopropyl)benzene


Formula: BrH₂CCH₂CH₂(C₆H₅)
MW: 199.09 g/mol
Boiling Pt: 219…220 °C
Density: 1.31 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000257
CAS Number: 637-59-2
EINECS: 211-294-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty