Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-hydroxybenzoic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR10133-100G OR10133-25G
76561-368EA 51 USD InStock
76561-368 76561-338
2-Amino-5-hydroxybenzoic acid 95%
2-Amino-5-hydroxybenzoic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 247 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007870
CAS Number: 394-31-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty