Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Thiophenedicarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Thiophene-2,5-dicarboxylic acid

OR10148-100G OR10148-500G OR10148-50G
76563-210EA 13 USD InStock
76563-210 76562-798 76562-746
2,5-Thiophenedicarboxylic acid
2,5-Thiophenedicarboxylic acid


Formula: C₆H₄O₄S
MW: 172.16 g/mol
Melting Pt: 355…360 °C
Storage Temperature: Ambient
MDL Number: MFCD00016896
CAS Number: 4282-31-9
EINECS: 224-284-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty