Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Nitroindole

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 6-Nitro-1H-indole

OR10187-5G OR10187-1G
76561-280EA 18.45 USD InStock
76561-280 76562-832
6-Nitroindole
6-Nitroindole


Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 137…143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051497
CAS Number: 4769-96-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty