Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-2-bromothiazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Ethyl 2-bromothiazole-4-carboxylate

OR10474-1G OR10474-5G
76560-514EA 13.33 USD InStock
76560-514 76560-202
Ethyl-2-bromothiazole-4-carboxylate
Ethyl-2-bromothiazole-4-carboxylate


Formula: C₆H₆BrNO₂S
MW: 236.08 g/mol
Melting Pt: 68…70 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788564
CAS Number: 100367-77-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty