Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydropyran-4-carboxylic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Tetrahydropyran-4-yl-carboxylic acid

OR10587-25G OR10587-100G OR10587-5G
76563-064EA 28 USD InStock
76563-064 76562-910 76561-640
Tetrahydropyran-4-carboxylic acid 98%
Tetrahydropyran-4-carboxylic acid


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00031016
CAS Number: 5337-03-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty