Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-4-bromoaniline

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Bromoaniline, N-BOC protectedtert-Butyl (4-bromophenyl)carbamate

OR10620-1G OR10620-5G OR10620-25G
76562-696EA 51 USD InStock
76562-696 76560-830 76561-504
N-Boc-4-bromoaniline
N-Boc-4-bromoaniline


Formula: C₁₁H₁₄BrNO₂
MW: 272.14 g/mol
Melting Pt: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD01006612
CAS Number: 131818-17-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty