Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloroimidazo[1,2-a]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR110406-1G
76564-094EA 71 USD InStock
76564-094
3-Chloroimidazo[1,2-a]pyridine
3-Chloroimidazo[1,2-a]pyridine


Formula: C₇H₅ClN₂
MW: 152.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07778355
CAS Number: 5315-73-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty