Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',4'-Dichloroacetophenone

Supplier: APOLLO SCIENTIFIC
Danger

OR11049-100G OR11049-10G OR11049-250G
76564-208EA 13.39 USD InStock
76564-208 76564-210 76564-212
2',4'-Dichloroacetophenone
2',4'-Dichloroacetophenone


Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 245…247 °C
Melting Pt: 31…34 °C
Density: 1.32 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000581
CAS Number: 2234-16-4
EINECS: 218-780-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty