Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR110710-250MG OR110710-5G OR110710-1G
76564-294EA 18.45 USD InStock
76564-294 76564-296 76564-298
Ethyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate
Ethyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate


Formula: C₁₀H₉BrN₂O₂
MW: 269.10
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD12827886
CAS Number: 1215504-30-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty