Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-nitrotoluene 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR11140-25G OR11140-100G OR11140-10G
76564-816EA 45 USD InStock
76564-816 76564-812 76564-814
4-Bromo-2-nitrotoluene 96%
4-Bromo-2-nitrotoluene


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 130 °C/12mmHg
Melting Pt: 45…48 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041243
CAS Number: 60956-26-5
EINECS: 262-536-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty