Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methoxybenzimidazole

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-Methoxy-1H-benzo[d]imidazole

OR11201-5G OR11201-1G OR11201-10G
76564-990EA 77 USD InStock
76564-990 76564-988 76564-986
5-Methoxybenzimidazole
5-Methoxybenzimidazole


Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 121…126 …lit °C
Storage Temperature: Ambient
MDL Number: MFCD00272526
CAS Number: 4887-80-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty