Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methylisophthalic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR12006-5G OR12006-100G OR12006-25G
76565-528EA 17 USD InStock
76565-528 76565-526 76565-530
5-Methylisophthalic acid
5-Methylisophthalic acid


Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 299…303 °C
Storage Temperature: Ambient
MDL Number: MFCD00013986
CAS Number: 499-49-0
EINECS: 207-881-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty