Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrobenzaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR12008-500G OR12008-100G
76565-538EA 130 USD InStock
76565-538 76565-536
4-Nitrobenzaldehyde 95%
4-Nitrobenzaldehyde


Formula: C₇H₅NO₃
MW: 151.12 g/mol
Melting Pt: 103…106 °C
Density: 1.496 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007346
CAS Number: 555-16-8
EINECS: 209-084-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty