Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-chlorobenzenesulfonic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR12580-100MG
76565-802EA 13.33 USD InStock
76565-802
2-Amino-5-chlorobenzenesulfonic acid
2-Amino-5-chlorobenzenesulphonic acid


Formula: C₆H₆ClNO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00035776
CAS Number: 133-74-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty