Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Homovanillic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-(4-Hydroxy-3-methoxyphenyl)acetic acid(4-Hydroxy-3-methoxyphenyl)acetic acid

OR13029-25G OR13029-5G
76566-030EA 52 USD InStock
76566-030 76566-032
Homovanillic acid 99%
Homovanillic acid


Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
MW: 182.18 g/mol
Melting Pt: 140…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00004350
CAS Number: 306-08-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty