Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-2-methylpyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR13032-5G OR13032-25G OR13032-1G
76566-050EA 51 USD InStock
76566-050 76566-046 76566-048
3-Bromo-2-methylpyridine
3-Bromo-2-methylpyridine


Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 65…67 °C/10mmHg
Density: 1.495 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00191224
CAS Number: 38749-79-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty