Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-ethylbenzene

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-(Bromophenyl)ethane

OR13057-100G OR13057-25G OR13057-500G
76566-084EA 13.33 USD InStock
76566-084 76566-086 76566-088
1-Bromo-4-ethylbenzene
1-Bromo-4-ethylbenzene


Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 203…204 °C
Density: 1.341 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00000111
CAS Number: 1585-07-5
EINECS: 216-439-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty