Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2-bromopyridine

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2-Bromopyridin-4-amine2-Bromo-4-pyridylamine

OR13064-25G OR13064-5G OR13064-100G
76566-108EA 19 USD InStock
76566-108 76566-110 76566-112
4-Amino-2-bromopyridine
4-Amino-2-bromopyridine


Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 92…99 °C
Storage Temperature: Ambient
MDL Number: MFCD01646061
CAS Number: 7598-35-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty