Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Benzyloxyindole

Supplier: APOLLO SCIENTIFIC
Warning

OR13129-25G OR13129-5G OR13129-1G
76566-160EA 42 USD InStock
76566-160 76566-162 76566-158
4-Benzyloxyindole
4-Benzyloxyindole


Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047200
CAS Number: 20289-26-3
EINECS: 243-690-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty