Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2-amino-5-chlorobenzoate

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Methyl 2-amino-5-chlorobenzoate

OR13133-25G OR13133-100G OR13133-5G
76566-170EA 51 USD InStock
76566-170 76566-172 76566-168
Methyl-2-amino-5-chlorobenzoate
Methyl 2-amino-5-chlorobenzoate


Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 168…170 °C/22mmHg
Melting Pt: 66…70 °C
Storage Temperature: Ambient
EINECS: 225-992-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty