Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxybenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR13187-1KG OR13187-500G OR13187-5KG
76566-206EA 41 USD InStock
76566-206 76566-208 76566-204
4-Hydroxybenzaldehyde
4-Hydroxybenzaldehyde


Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 191 °C/50mmHg
Melting Pt: 112…116 °C
Density: 1.129 g/cm³ (25 °C)
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00006939
CAS Number: 123-08-0
EINECS: 204-599-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty