Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-chloropyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-Chloropyridin-2-amine5-Chloro-2-pyridylamine

OR13201-100G OR13201-5G OR13201-25G
76566-232EA 11.33 USD InStock
76566-232 76566-234 76566-236
2-Amino-5-chloropyridine 98%
2-Amino-5-chloropyridine


Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 127…128 °C/11mmHg
Melting Pt: 135…138 °C
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00006324
CAS Number: 1072-98-6
EINECS: 214-020-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty