Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-4-Chloromandelic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Chloromandelic acid(±)-4-Chlorophenylglycolic acid

OR13218-25G OR13218-5G OR13218-100G
76566-240EA 13.33 USD InStock
76566-240 76566-242 76566-244
(±)-4-Chloromandelic acid
(±)-4-Chloromandelic acid


Formula: C₈H₇ClO₃
MW: 186.59 g/mol
Melting Pt: 117…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00042724
CAS Number: 492-86-4
EINECS: 207-764-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty