Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-methyl-1H-1,2,3-triazole-4-carboxamide

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-(4-Amino-1,2,5-oxadiazol-3-yl)-4-carbamoyl-5-methyl-1H-1,2,3-triazole3-Amino-4-(4-carbamoyl-5-methyl-1H-1,2,3-triazol-1-yl)-1,2,5-oxadiazole

OR13280-100MG
76566-278EA 80 USD InStock
76566-278
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-methyl-1H-1,2,3-triazole-4-carboxamide
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-methyl-1H-1,2,3-triazole-4-carboxamide


Formula: C₆H₇N₇O₂
MW: 209.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01240850
CAS Number: 312267-71-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty