Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(Aminomethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-(1,2,3,4-Tetrahydroisoquinolin-1-yl)methylamine dihydrochloride

OR13424-250MG OR13424-100MG OR13424-1G
76566-316EA 499 USD InStock
76566-316 76566-312 76566-314
1-(Aminomethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
1-(Aminomethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride


Formula: C₁₀H₁₄N₂·2ClH
MW: 235.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00227022
CAS Number: 140845-50-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty