Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5,7-Dimethylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Carboxy-5,7-dimethylpyrazolo[1,5-a]pyrimidine

OR13446-250MG OR13446-1G
76566-344EA 47.38 USD InStock
76566-344 76566-346
5,7-Dimethylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid
5,7-Dimethylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid


Formula: C₉H₉N₃O₂
MW: 191.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00461394
CAS Number: 300691-07-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty