Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-2-chloropyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Chloropyridin-5-amine2-Chloro-5-pyridylamine6-Chloropyridin-3-amine

OR1348-25G OR1348-1G OR1348-5G
76566-360EA 17 USD InStock
76566-360 76566-356 76566-358
5-Amino-2-chloropyridine 98%
5-Amino-2-chloropyridine


Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00006243
CAS Number: 5350-93-6
EINECS: 226-322-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty